missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Adamantyl methyl ketone, 99%

Manufacturer:  Alfa Aesar™ H5588906

 View more versions of this product

Catalog No. AAH5588906



Description

Description

Applications
1-Adamantyl methyl ketone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol- very faint turbidity. Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
Specifications

Specifications

1-Adamantyl methyl ketone
53°C to 55°C
1660-04-4
MFCD00074739
Soluble in methanol- very faint turbidity. Insoluble in water.
CC(=O)C12CC3CC(C1)CC(C3)C2
178.275
178.27
108°C (226°F)
5g
C12H18O
1-1-adamantyl ethanone
DACIGVIOAFXPHW-UHFFFAOYSA-N
1-(1-adamantyl)ethanone
123126
99%

Chemical Identifiers

1660-04-4
178.275
DACIGVIOAFXPHW-UHFFFAOYSA-N
123126
CC(=O)C12CC3CC(C1)CC(C3)C2
C12H18O
MFCD00074739
1-1-adamantyl ethanone
1-(1-adamantyl)ethanone
Safety and Handling

Safety and Handling

EINECSNumber : 216-761-9

TSCA : No

Recommended Storage : Ambient temperatures

Documents