missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Adamantyl methyl ketone, 99%, Thermo Scientific Chemicals
$52.05 - $169.05
Chemical Identifiers
CAS | 1660-04-4 |
---|---|
Molecular Formula | C12H18O |
Molecular Weight (g/mol) | 178.28 |
MDL Number | MFCD00074739 |
InChI Key | DACIGVIOAFXPHW-UHFFFAOYSA-N |
Synonym | 1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone |
PubChem CID | 123126 |
IUPAC Name | 1-(1-adamantyl)ethanone |
SMILES | CC(=O)C12CC3CC(CC(C3)C1)C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5588903
|
Thermo Scientific Chemicals
H5588903 |
1 g |
N/A
|
|
|||||
AAH5588906
|
Thermo Scientific Chemicals
H5588906 |
5 g |
N/A
|
|
|||||
Description
1-Adamantyl methyl ketone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Adamantyl methyl ketone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol- very faint turbidity. Insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
Chemical Identifiers
1660-04-4 | |
178.28 | |
DACIGVIOAFXPHW-UHFFFAOYSA-N | |
123126 | |
CC(=O)C12CC3CC(CC(C3)C1)C2 |
C12H18O | |
MFCD00074739 | |
1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone | |
1-(1-adamantyl)ethanone |
Specifications
1660-04-4 | |
108°C (226°F) | |
MFCD00074739 | |
1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone | |
DACIGVIOAFXPHW-UHFFFAOYSA-N | |
1-(1-adamantyl)ethanone | |
123126 | |
99% |
53°C to 55°C | |
C12H18O | |
1 g | |
Soluble in methanol- very faint turbidity. Insoluble in water. | |
CC(=O)C12CC3CC(CC(C3)C1)C2 | |
178.28 | |
178.27 | |
1-Adamantyl methyl ketone |
Safety and Handling
EINECSNumber : 216-761-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only