missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Adamantanol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 157100050

 View more versions of this product

Catalog No. AC157100050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Adamantanol
Glass bottle
>240.0°C
768-95-6
100.0
C10H16O
1-adamantanol
VLLNJDMHDJRNFK-UHFFFAOYSA-N
adamantan-1-ol
64152
Crystalline Powder or Needles
Authentic
White
5g
98.5
98.5% min. (GC)
MFCD00074729
Solubility in water: insoluble in water. Other solubilities: soluble in ethanol,diethyl ether,methylene,chloride and acetone
C1C2CC3CC1CC(C2)(C3)O
152.24
152.24
99%

Chemical Identifiers

768-95-6
152.24
VLLNJDMHDJRNFK-UHFFFAOYSA-N
64152
C1C2CC3CC1CC(C2)(C3)O
C10H16O
MFCD00074729
1-adamantanol
adamantan-1-ol
Safety and Handling

Safety and Handling

EINECSNumber : 212-202-8

SDS Documents