missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Adamantanol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1020918
Description
1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.
Solubility
Soluble in ethanol , methanol, and diethyl ether. Partly miscible in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
1-Adamantanol | |
∼245°C (sublimation) | |
MFCD00074729 | |
1903952 | |
Soluble in ethanol ,methanol,and diethyl ether. Partly miscible in water. | |
C1C2CC3CC1CC(C2)(C3)O | |
152.237 | |
152.24 |
768-95-6 | |
C10H16O | |
50 g | |
1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol | |
VLLNJDMHDJRNFK-UHFFFAOYSA-N | |
adamantan-1-ol | |
64152 | |
99% |
Chemical Identifiers
768-95-6 | |
152.237 | |
VLLNJDMHDJRNFK-UHFFFAOYSA-N | |
64152 | |
C1C2CC3CC1CC(C2)(C3)O |
C10H16O | |
MFCD00074729 | |
1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol | |
adamantan-1-ol |
Safety and Handling
EINECSNumber : 212-202-8
RTECSNumber : AU4980000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only