missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Adamantanol, 99%

Manufacturer:  Alfa Aesar™ A1020930

 View more versions of this product

Catalog No. AAA1020930



Description

Description

Applications
1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.

Solubility
Soluble in ethanol , methanol, and diethyl ether. Partly miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications

Specifications

1-Adamantanol
250g
C10H16O
1903952
VLLNJDMHDJRNFK-UHFFFAOYSA-N
adamantan-1-ol
64152
99%
~245°C (sublimation)
768-95-6
MFCD00074729
1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1~3,7~ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol
C1C2CC3CC1CC(C2)(C3)O
152.237
152.24

Chemical Identifiers

768-95-6
152.237
VLLNJDMHDJRNFK-UHFFFAOYSA-N
64152
C1C2CC3CC1CC(C2)(C3)O
C10H16O
MFCD00074729
1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1~3,7~ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol
adamantan-1-ol
Safety and Handling

Safety and Handling

RTECSNumber : AU4980000

SDS Documents