Learn More
1-Adamantanemethylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6619706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Adamantanemethylamine | |
Colorless to Yellow | |
83°C to 85°C (0.3 mmHg) | |
C11H20N | |
MFCD00074750 | |
UN2735 | |
1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine | |
[NH3+]CC12CC3CC(CC(C3)C1)C2 | |
166.29 | |
168.28 | |
Liquid |
17768-41-1 | |
0.933 g/mL | |
92°C (198°F) | |
1.512 | |
5 g | |
Air Sensitive | |
XSOHXMFFSKTSIT-UHFFFAOYSA-O | |
(adamantan-1-yl)methanaminium | |
86625 | |
97% |
Chemical Identifiers
17768-41-1 | |
166.29 | |
XSOHXMFFSKTSIT-UHFFFAOYSA-O | |
86625 | |
[NH3+]CC12CC3CC(CC(C3)C1)C2 |
C11H20N | |
MFCD00074750 | |
1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine | |
(adamantan-1-yl)methanaminium |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 241-752-1
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only