Learn More
1-Adamantanamine, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 160470050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.
Solubility
Soluble in organic solvents. Insoluble in water.
Notes
Use only in a chemical fume hood. Keep away from sources of ignition. Incompatible with oxidizing agent.
Specifications
1-Adamantanamine | |
95 | |
208.0°C | |
Authentic | |
Glass bottle | |
MFCD00074732 | |
15, 368 | |
Solubility in water: sparingly soluble | |
C1C2CC3CC1CC(C2)(C3)N | |
151.25 | |
CHEBI:2618 | |
96% |
768-94-5 | |
100 | |
White to Cream | |
0.96 | |
C10H17N | |
5 g | |
amantadine, 1-adamantanamine, 1-adamantylamine, 1-aminoadamantane, adamantanamine, adamantylamine, aminoadamantane, amantidine, symadine, symmetrel | |
DKNWSYNQZKUICI-UHFFFAOYSA-N | |
adamantan-1-amine | |
2130 | |
151.25 | |
Powder |
Chemical Identifiers
768-94-5 | |
151.25 | |
DKNWSYNQZKUICI-UHFFFAOYSA-N | |
2130 | |
adamantan-1-amine |
C10H17N | |
MFCD00074732 | |
amantadine, 1-adamantanamine, 1-adamantylamine, 1-aminoadamantane, adamantanamine, adamantylamine, aminoadamantane, amantidine, symadine, symmetrel | |
CHEBI:2618 | |
C1C2CC3CC1CC(C2)(C3)N |
Safety and Handling
Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, Causes skin irritation
Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
EINECSNumber : 212-201-2
RUO – Research Use Only