missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acryloyloxy-3-hydroxyadamantane 98.0+%, TCI America™
Supplier: TCI America A285925G
Spécifications
| 1-Acryloyloxy-3-hydroxyadamantane | |
| 72°C | |
| C13H18O3 | |
| 25 g | |
| DKDKCSYKDZNMMA-UHFFFAOYSA-N | |
| (3-hydroxy-1-adamantyl) prop-2-enoate | |
| 22228790 | |
| ≥98.0% (GC) |
| 216581-76-9 | |
| White | |
| MFCD13194929 | |
| 3-Hydroxy-1-adamantyl Acrylate, Acrylic Acid 3-Hydroxy-1-adamantyl Ester | |
| C=CC(=O)OC12CC3CC(C1)CC(C3)(C2)O | |
| 222.284 | |
| 222.28 | |
| Crystalline Powder |
Identifiants chimiques
| 216581-76-9 | |
| 222.284 | |
| DKDKCSYKDZNMMA-UHFFFAOYSA-N | |
| 22228790 | |
| C=CC(=O)OC12CC3CC(C1)CC(C3)(C2)O |
| C13H18O3 | |
| MFCD13194929 | |
| 3-Hydroxy-1-adamantyl Acrylate, Acrylic Acid 3-Hydroxy-1-adamantyl Ester | |
| (3-hydroxy-1-adamantyl) prop-2-enoate |
Sécurité et manipulation
missing translation for 'einecsNumber' : (4)-1910
missing translation for 'tsca' : Yes