Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetylpyrene, 97%

$79.24 - $79.24

Chemical Identifiers

Chemical Name or Material 1-Acetylpyrene
CAS 3264-21-9
Molecular Formula C18H12O
Formula Weight 244.29
MDL Number MFCD00021663
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2248388

  SDS
alfa aesar™
B2248388
2.5g Each for $79.24
Description & Specifications

Specifications

Formula Weight 244.29
CAS 3264-21-9
Melting Point 86° to 89°C
Quantity 2.5g
Chemical Name or Material 1-Acetylpyrene
MDL Number MFCD00021663
Molecular Formula C18H12O
Assay 97%