Please login to your online account to display your discounted pricing

1-Acetylpiperidine-4-carbonitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Acetylpiperidine-4-carbonitrile
Assay Percent Range 97%
Molecular Formula C8H12N2O
Formula Weight 152.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW04588CB

 
fisher scientific company
SEW04588CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.2
Quantity 250mg
Chemical Name or Material 1-Acetylpiperidine-4-carbonitrile
Assay Percent Range 97%
Molecular Formula C8H12N2O
CAS 25503-91-7