Please login to your online account to display your discounted pricing

1-Acetylpiperidine-4-carbonitrile, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  SEW04588CB

 View more versions of this product

Catalog No. SEW04588CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 152.2
Quantity 250mg
Molecular Formula C8H12N2O
CAS 25503-91-7
Chemical Name or Material 1-Acetylpiperidine-4-carbonitrile
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-Acetylpiperidine-4-carbonitrile
Assay Percent Range 97%
Molecular Formula C8H12N2O
Formula Weight 152.2