Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetylindole-3-carboxaldehyde, 98%

$25.76 - $320.95

Chemical Identifiers

Chemical Name or Material 1-Acetylindole-3-carboxaldehyde
CAS 22948-94-3
Molecular Formula C11H9NO2
Formula Weight 187.2
MDL Number MFCD00039691
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2152703

  SDS
alfa aesar™
B2152703
1g Each for $25.76

AAB2152714

  SDS
alfa aesar™
B2152714
25g Each for $320.95

AAB2152706

  SDS
alfa aesar™
B2152706
5g Each for $78.80
Description & Specifications

Specifications

Formula Weight 187.2
Molecular Formula C11H9NO2
Health Hazard 1 Warning
MDL Number MFCD00039691
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
CAS 22948-94-3
Quantity 1g
Chemical Name or Material 1-Acetylindole-3-carboxaldehyde
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 160° to 164°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.