missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetylimidazole, 98%

$26.22 - $137.70

Chemical Identifiers

CAS 2466-76-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD00005287
InChI Key VIHYIVKEECZGOU-UHFFFAOYSA-N
Synonym 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
PubChem CID 17174
ChEBI CHEBI:16984
IUPAC Name 1-imidazol-1-ylethanone
SMILES CC(=O)N1C=CN=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0013706 View Documents Alfa Aesar™
L0013706
5g Each for $26.22
 
AAL0013722 View Documents Alfa Aesar™
L0013722
100g Each for $137.70
 
Description

Description

Applications
It is a mild acylating agent and its reactivity can be enhanced by quaternization with, e.g. benzyl bromide. 1-Acetylimidazole was used as acetylation reagent for amino groups. It was also employed for acetylation of histones. It is relatively specific reagent for tyrosyl residue acetylatioa dn a reagent used in the synthesis of annulated imidazole derivatives.

Solubility
Hydrolyzes in water. Solubility in methanol (almost transparency).

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Chemical Identifiers

2466-76-4
110.116
VIHYIVKEECZGOU-UHFFFAOYSA-N
17174
1-imidazol-1-ylethanone
C5H6N2O
MFCD00005287
1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
CHEBI:16984
CC(=O)N1C=CN=C1

Specifications

98°C to 104°C
2466-76-4
MFCD00005287
1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
CC(=O)N1C=CN=C1
110.116
CHEBI:16984
98%
5g
C5H6N2O
108425
VIHYIVKEECZGOU-UHFFFAOYSA-N
1-imidazol-1-ylethanone
17174
110.12
1-Acetylimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NI3400000

TSCA : TSCA