missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetylimidazole, 98%

Manufacturer:  Alfa Aesar™ L0013722

 View more versions of this product

Catalog No. AAL0013722



Description

Description

Applications
It is a mild acylating agent and its reactivity can be enhanced by quaternization with, e.g. benzyl bromide. 1-Acetylimidazole was used as acetylation reagent for amino groups. It was also employed for acetylation of histones. It is relatively specific reagent for tyrosyl residue acetylatioa dn a reagent used in the synthesis of annulated imidazole derivatives.

Solubility
Hydrolyzes in water. Solubility in methanol (almost transparency).

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Specifications

1-Acetylimidazole
98°C to 104°C
2466-76-4
MFCD00005287
1-1h-imidazol-1-yl ethanone, 1-acetyl-1h-imidazole, 1-acetylimidazole, 1h-imidazole, 1-acetyl, 3-acetylimidazole, acetylimidazole, ccris 6534, imidazole, 1-acetyl, n-acetylimidazole, unii-zmp8x1y11g
VIHYIVKEECZGOU-UHFFFAOYSA-N
1-imidazol-1-ylethanone
17174
110.12
Light and moisture sensitive
100g
C5H6N2O
108425
Hydrolyzes in water. Solubility in methanol (almost transparency).
CC(=O)N1C=CN=C1
110.116
CHEBI:16984
98%

Chemical Identifiers

2466-76-4
110.116
VIHYIVKEECZGOU-UHFFFAOYSA-N
17174
1-imidazol-1-ylethanone
C5H6N2O
MFCD00005287
1-1h-imidazol-1-yl ethanone, 1-acetyl-1h-imidazole, 1-acetylimidazole, 1h-imidazole, 1-acetyl, 3-acetylimidazole, acetylimidazole, ccris 6534, imidazole, 1-acetyl, n-acetylimidazole, unii-zmp8x1y11g
CHEBI:16984
CC(=O)N1C=CN=C1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 219-577-7

RTECSNumber : NI3400000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents