Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetylhomopiperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-Acetylhomopiperazine
CAS 61903-11-5
Molecular Formula C7H14N2O
Formula Weight 142.2
MDL Number MFCD00674492
UN Number UN2735
Synonym 1-Acetyl-1,4-diazepane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5026003

  SDS
alfa aesar™
H5026003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50260MD

  SDS
alfa aesar™
H50260MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.2
Chemical Name or Material 1-Acetylhomopiperazine
Assay 97%
Molecular Formula C7H14N2O
MDL Number MFCD00674492
CAS 61903-11-5
UN Number UN2735
Synonym 1-Acetyl-1,4-diazepane
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.