Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetylferrocene, 99+%

Chemical Identifiers

Chemical Name or Material 1-Acetylferrocene
CAS 1271-55-2
Formula Weight 228.07g/mol
MDL Number MFCD00001432
UN Number UN3467
Synonym Acetylferrocene; [(Acetylcyclopentadienyl)cyclopentadienyliron]
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2295406

 
alfa aesar™
2295406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Melting Point 81° to 83°C
Quantity 5g
Chemical Name or Material 1-Acetylferrocene
CAS 1271-55-2
Physical Form Crystalline
Assay ≥99%
Formula Weight 228.07g/mol
MDL Number MFCD00001432
UN Number UN3467
Synonym Acetylferrocene; [(Acetylcyclopentadienyl)cyclopentadienyliron]
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310
Fatal if swallowed.
Fatal in contact with skin.
Health Hazard 3 GHS P Statement
P280-P301+P310-P361-P302+P350-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P361-P302+P350-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310
Fatal if swallowed.
Fatal in contact with skin.