Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetylferrocene, 97%

Chemical Identifiers

Chemical Name or Material 1-Acetylferrocene
CAS 1271-55-2
Molecular Formula C12H12FeO
Formula Weight 228.07g/mol
MDL Number MFCD00001432
UN Number UN3467
Synonym (Acetylcyclopentadienyl)cyclopentadienyliron; Ferrocenyl methyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1307806

 
alfa aesar™
A13078-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1307822

 
alfa aesar™
A13078-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1307814

 
alfa aesar™
A13078-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 160° to 163°C (4mmHg)
Melting Point 79° to 86°C
Quantity 5g
Chemical Name or Material 1-Acetylferrocene
CAS 1271-55-2
Assay 97%
Molecular Formula C12H12FeO
Formula Weight 228.07g/mol
MDL Number MFCD00001432
UN Number UN3467
Synonym (Acetylcyclopentadienyl)cyclopentadienyliron; Ferrocenyl methyl ketone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310
Fatal if swallowed.
Fatal in contact with skin.
Health Hazard 3 GHS P Statement
P280-P301+P310-P361-P302+P350-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
RTECS Number OB3700000
TSCA TSCA

Applications
1-Acetylferrocene was originally used in military or space field as an additive in rocket propellant, to promote the burning rate. Its ferrocenyl derivative has wide applications to biological and medical fields such as ferrocene-modified beta-lactam because of its physiological activity of anti-malarial, anti-tumor, bactericidal, anti-inflammatory, treatment of anemia, inhibition of enzymatic activity and so on by virtue of its unique structure and diverse properties.

Solubility
Insoluble in water. Soluble in most organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P361-P302+P350-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310
Fatal if swallowed.
Fatal in contact with skin.