Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetyl-5-nitroindoline, 98%

$197.03 - $803.50

Chemical Identifiers

Chemical Name or Material 1-Acetyl-5-nitroindoline
CAS 33632-27-8
Molecular Formula C10H10N2O3
Formula Weight 206.2
MDL Number MFCD00005811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2021209

 
alfa aesar™
B2021209
10g Each for $197.03

AAB2021218

 
alfa aesar™
B2021218
50g Each for $803.50
Description & Specifications

Specifications

Formula Weight 206.2
Chemical Name or Material 1-Acetyl-5-nitroindoline
TSCA TSCA
Assay 98%
Molecular Formula C10H10N2O3
MDL Number MFCD00005811
CAS 33632-27-8
Melting Point 175° to 178°C
Quantity 10g
RTECS Number NM1892800