missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetyl-5-bromo-7-nitroindoline, 98%

$44.86 - $133.85

Chemical Identifiers

CAS 62368-07-4
Molecular Formula C10H9BrN2O3
Molecular Weight (g/mol) 285.097
MDL Number MFCD00056018
InChI Key RCELVCGNAKOBPO-UHFFFAOYSA-N
Synonym 1-acetyl-5-bromo-7-nitroindoline, 1-5-bromo-7-nitroindolin-1-yl ethanone, n-acetyl-5-bromo-7-nitro-indoline, 1-acetyl-5-bromo-7-nitroindole, 1-5-bromo-7-nitro-2,3-dihydroindol-1-yl ethanone, 1-acetyl-5-bromo-7-nitro-2,3-dihydro-1h-indole, pubchem7303, acmc-1bfxp, n-acetyl-5-bromo-7-nitroindoline, 1-acetyl-5-bromo-7-nitroindoline
PubChem CID 2729243
IUPAC Name 1-(5-bromo-7-nitro-2,3-dihydroindol-1-yl)ethanone
SMILES CC(=O)N1CCC2=CC(=CC(=C21)[N+](=O)[O-])Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1827103 View Documents Alfa Aesar™
L1827103
1g Each for $44.86
 
AAL1827106 View Documents Alfa Aesar™
L1827106
5g Each for $133.85
 
Specifications

Chemical Identifiers

62368-07-4
285.097
RCELVCGNAKOBPO-UHFFFAOYSA-N
2729243
CC(=O)N1CCC2=CC(=CC(=C21)[N+](=O)[O-])Br
C10H9BrN2O3
MFCD00056018
1-acetyl-5-bromo-7-nitroindoline, 1-5-bromo-7-nitroindolin-1-yl ethanone, n-acetyl-5-bromo-7-nitro-indoline, 1-acetyl-5-bromo-7-nitroindole, 1-5-bromo-7-nitro-2,3-dihydroindol-1-yl ethanone, 1-acetyl-5-bromo-7-nitro-2,3-dihydro-1h-indole, pubchem7303, acmc-1bfxp, n-acetyl-5-bromo-7-nitroindoline, 1-acetyl-5-bromo-7-nitroindoline
1-(5-bromo-7-nitro-2,3-dihydroindol-1-yl)ethanone

Specifications

196°C to 198°C
62368-07-4
MFCD00056018
1-acetyl-5-bromo-7-nitroindoline, 1-5-bromo-7-nitroindolin-1-yl ethanone, n-acetyl-5-bromo-7-nitro-indoline, 1-acetyl-5-bromo-7-nitroindole, 1-5-bromo-7-nitro-2,3-dihydroindol-1-yl ethanone, 1-acetyl-5-bromo-7-nitro-2,3-dihydro-1h-indole, pubchem7303, acmc-1bfxp, n-acetyl-5-bromo-7-nitroindoline, 1-acetyl-5-bromo-7-nitroindoline
CC(=O)N1CCC2=CC(=CC(=C21)[N+](=O)[O-])Br
285.097
285.1
1-Acetyl-5-bromo-7-nitroindoline
1g
C10H9BrN2O3
237670
RCELVCGNAKOBPO-UHFFFAOYSA-N
1-(5-bromo-7-nitro-2,3-dihydroindol-1-yl)ethanone
2729243
98%
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger