Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetyl-4-piperidone, 99%

$58.77 - $219.99

Chemical Identifiers

Chemical Name or Material 1-Acetyl-4-piperidone
CAS 32161-06-1
Molecular Formula C7H11NO2
Formula Weight 141.17
MDL Number MFCD00006190
Beilstein 116031
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1351206

  SDS
alfa aesar™
A1351206
5g Each for $58.77

AAA1351214

  SDS
alfa aesar™
A1351214
25g Each for $219.99
Description & Specifications

Specifications

Formula Weight 141.17
Assay 99%
CAS 32161-06-1
Beilstein 116031
Quantity 5g
Boiling Point 218°C
Chemical Name or Material 1-Acetyl-4-piperidone
MDL Number MFCD00006190
Flash Point 112°C (234°F)
Molecular Formula C7H11NO2

Applications
1-Acetyl-4-piperidone is used as an intermediate in the manufacture of chemicals and pharmaceutical drugs, and manufacture of fentanyl.

Solubility
Soluble in water. Insoluble in acetic acid.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.