missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Acetyl-4-piperidone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1351206
Description
1-Acetyl-4-piperidone is used as an intermediate in the manufacture of chemicals and pharmaceutical drugs, and manufacture of fentanyl.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Acetyl-4-piperidone is used as an intermediate in the manufacture of chemicals and pharmaceutical drugs, and manufacture of fentanyl.
Solubility
Soluble in water. Insoluble in acetic acid.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
1-Acetyl-4-piperidone | |
1.146 | |
112°C (234°F) | |
1.503 | |
5 g | |
1-acetyl-4-piperidone, n-acetyl-4-piperidone, 4-piperidinone, 1-acetyl, 1-acetyl-4-piperidinone, 1-acetyl-piperidin-4-one, 1-acetyl-4-oxopiperidine, n-acetyl-4-piperidinone, n-acetyl4-piperidone, pubchem19615, 1-acetylpiperid-4-one | |
NNFOVLFUGLWWCL-UHFFFAOYSA-N | |
1-acetylpiperidin-4-one | |
122563 | |
99% |
32161-06-1 | |
218°C | |
C7H11NO2 | |
MFCD00006190 | |
116031 | |
Soluble in water. Insoluble in acetic acid. | |
CC(=O)N1CCC(=O)CC1 | |
141.17 | |
141.17 |
Chemical Identifiers
32161-06-1 | |
141.17 | |
NNFOVLFUGLWWCL-UHFFFAOYSA-N | |
122563 | |
CC(=O)N1CCC(=O)CC1 |
C7H11NO2 | |
MFCD00006190 | |
1-acetyl-4-piperidone, n-acetyl-4-piperidone, 4-piperidinone, 1-acetyl, 1-acetyl-4-piperidinone, 1-acetyl-piperidin-4-one, 1-acetyl-4-oxopiperidine, n-acetyl-4-piperidinone, n-acetyl4-piperidone, pubchem19615, 1-acetylpiperid-4-one | |
1-acetylpiperidin-4-one |
Safety and Handling
EINECSNumber : 250-937-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only