Please login to your online account to display your discounted pricing

1-Acetyl-4-methylpiperazine hydrochloride, Tocris

Nicotinic agonist

$162.94 - $684.37

Chemical Identifiers

Chemical Name or Material 1-Acetyl-4-methylpiperazine hydrochloride
Assay Percent Range >98%
Molecular Formula C7H14N2O·HCl
Formula Weight 178.66
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

035110

 
r & d systems
035110
10mg Each for $162.94

035150

 
r & d systems
035150
50mg Each for $684.37
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 178.66
Assay Percent Range >98%
Molecular Formula C7H14N2O·HCl
Chemical Name or Material 1-Acetyl-4-methylpiperazine hydrochloride
CAS 144205-68-5
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature