Please login to your online account to display your discounted pricing

1-Acetyl-4-methylpiperazine hydrochloride, Tocris


Nicotinic agonist

Quantity 50mg

Manufacturer: r & d systems  035150

 View more versions of this product

Catalog No. 035150

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 178.66
Assay Percent Range >98%
CAS 144205-68-5
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material 1-Acetyl-4-methylpiperazine hydrochloride
Molecular Formula C7H14N2O·HCl

Specifications

Chemical Name or Material 1-Acetyl-4-methylpiperazine hydrochloride
Assay Percent Range >98%
Molecular Formula C7H14N2O·HCl
Formula Weight 178.66
Accessories

Desiccate at Room Temperature