missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-(methylamino)piperidine

CAS: 139062-96-7 | C8H16N2O | 156.23 g/mol

Supplier:  Thermo Scientific Chemicals H5098503

 View more versions of this product

Catalog No. AAH5098503


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Acetyl-4-(methylamino)piperidine
C8H16N2O
1 g
1-acetyl-4-methylamino piperidine, 1-acetyl-n-methylpiperidin-4-amine, 1-4-methylamino piperidin-1-yl ethan-1-one, 1-4-methylamino piperidin-1-yl ethanone, 4-n-methylamino-1-acetyl-piperidine, 4-piperidinamine, 1-acetyl-n-methyl-9ci, 4-n-methylamino-1-acetyl, acmc-209vt3, 1-acetyl-4-methylaminopiperidine
CNC1CCN(CC1)C(C)=O
156.23
156.23
139062-96-7
MFCD07367773
UN3259
RSEPODZAQBVPOS-UHFFFAOYSA-N
1-[4-(methylamino)piperidin-1-yl]ethanone
19761265

Chemical Identifiers

139062-96-7
156.23
RSEPODZAQBVPOS-UHFFFAOYSA-N
19761265
C8H16N2O
MFCD07367773
1-acetyl-4-methylamino piperidine, 1-acetyl-n-methylpiperidin-4-amine, 1-4-methylamino piperidin-1-yl ethan-1-one, 1-4-methylamino piperidin-1-yl ethanone, 4-n-methylamino-1-acetyl-piperidine, 4-piperidinamine, 1-acetyl-n-methyl-9ci, 4-n-methylamino-1-acetyl, acmc-209vt3, 1-acetyl-4-methylaminopiperidine
CNC1CCN(CC1)C(C)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only