missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Acetyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one, 97%, Maybridge™

$103.46 - $103.46

Chemical Identifiers

CAS 34581-92-5
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00173863
InChI Key AINPDQRRTXHBAY-UHFFFAOYSA-N
Synonym 1-acetyl-4-methyl-1h-pyrrol-2 5h-one, 1-acetyl-4-methyl-2,5-dihydro-1h-pyrrol-2-one, 1-acetyl-4-methyl-5h-pyrrol-2-one, 2h-pyrrol-2-one,1-acetyl-1,5-dihydro-4-methyl
PubChem CID 2774997
IUPAC Name 1-acetyl-3-methyl-2H-pyrrol-5-one
SMILES CC1=CC(=O)N(C1)C(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB09313DA
SDS View Documents
Fisher Scientific Company
BTB09313DA
1g Amber Glass Bottle
Each for $103.46
 
Description

Description

Specifications

Chemical Identifiers

34581-92-5
139.154
AINPDQRRTXHBAY-UHFFFAOYSA-N
2774997
CC1=CC(=O)N(C1)C(=O)C
C7H9NO2
MFCD00173863
1-acetyl-4-methyl-1h-pyrrol-2 5h-one, 1-acetyl-4-methyl-2,5-dihydro-1h-pyrrol-2-one, 1-acetyl-4-methyl-5h-pyrrol-2-one, 2h-pyrrol-2-one,1-acetyl-1,5-dihydro-4-methyl
1-acetyl-3-methyl-2H-pyrrol-5-one

Specifications

Amber Glass Bottle
34581-92-5
100.0
C7H9NO2
1-acetyl-4-methyl-1h-pyrrol-2 5h-one, 1-acetyl-4-methyl-2,5-dihydro-1h-pyrrol-2-one, 1-acetyl-4-methyl-5h-pyrrol-2-one, 2h-pyrrol-2-one,1-acetyl-1,5-dihydro-4-methyl
CC1=CC(=O)N(C1)C(=O)C
139.154
139
1-Acetyl-4-methyl-2, 5-dihydro-1H-pyrrol-2-one
1g
96.5
97%
MFCD00173863
AINPDQRRTXHBAY-UHFFFAOYSA-N
1-acetyl-3-methyl-2H-pyrrol-5-one
2774997
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS