Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetyl-4-aminopiperidine hydrochloride, 97%

$254.22 - $254.22

Chemical Identifiers

Chemical Name or Material 1-Acetyl-4-aminopiperidine hydrochloride
CAS 214147-48-5
Molecular Formula C7H14N2O·HCl
Formula Weight 178.66g/mol
MDL Number MFCD08690151
Synonym 1-Acetyl-4-piperidinamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3038903

 
alfa aesar™
H30389-03
1g Each for $254.22
Description & Specifications

Specifications

Melting Point 231° to 234°C
Quantity 1g
Chemical Name or Material 1-Acetyl-4-aminopiperidine hydrochloride
CAS 214147-48-5
Assay 97%
Molecular Formula C7H14N2O·HCl
Formula Weight 178.66g/mol
MDL Number MFCD08690151
Synonym 1-Acetyl-4-piperidinamine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.