Please login to your online account to display your discounted pricing

1-Acetyl-4-(4-hydroxyphenyl)piperazine 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  100340500

 View more versions of this product

Catalog No. AC100340500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 220.27
Quantity 50g
Color White to slightly pink or beige
EINECS Number 267-744-8
Melting Point 180° to 185°C
CAS Max % 100.0
Total Impurity < 2% (TLC)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 98%
Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
CAS 67914-60-7
CAS Min % 97.5
MDL Number MFCD00044905
Molecular Formula C12H16N2O2
Transmittance 70% min. (400nm, 1cm cell) (10% min. in N, N-dimethylformamide)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
Name Note 98%
CAS 67914-60-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H16N2O2
Formula Weight 220.27
MDL Number MFCD00044905
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents