Please login to your online account to display your discounted pricing

1-Acetyl-4-(4-hydroxyphenyl)piperazine 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC100340250

 View more versions of this product

Catalog No. AC100340250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 220.27
Quantity 25g
Color White to slightly pink or beige
EINECS Number 267-744-8
Melting Point 180° to 185°C
CAS Max % 100.0
Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C12H16N2O2
MDL Number MFCD00044905
Total Impurity < 2% (TLC)
CAS 67914-60-7
Transmittance 70% min. (400nm, 1cm cell) (10% min. in N, N-dimethylformamide)
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
Name Note 98%
CAS 67914-60-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H16N2O2
Formula Weight 220.27
MDL Number MFCD00044905
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents