missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Acetyl-4-(4-hydroxyphenyl)piperazine 98%, ACROS Organics™

$80.99 - $80.99

Chemical Identifiers

CAS 67914-60-7
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD00044905
InChI Key AGVNLFCRZULMKK-UHFFFAOYSA-N
Synonym 1-acetyl-4-4-hydroxyphenyl piperazine, 4-1-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, n-acetyl-4-4-hydroxyphenyl piperazine, 4-4-acetyl-1-piperazinyl phenol, piperazine, 1-acetyl-4-4-hydroxyphenyl, 4-4-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethan-1-one, 1-acetyl-4-4-hydroxylphenyl piperazine
PubChem CID 712441
IUPAC Name 1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC100340500 SDS View Documents Acros Organics
100340500
50g Glass bottle Each for $80.99
AC100340250 SDS View Documents Acros Organics
AC100340250
25g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

67914-60-7
220.272
AGVNLFCRZULMKK-UHFFFAOYSA-N
712441
CC(=O)N1CCN(CC1)C2=CC=C(C=C2)O
C12H16N2O2
MFCD00044905
1-acetyl-4-4-hydroxyphenyl piperazine, 4-1-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, n-acetyl-4-4-hydroxyphenyl piperazine, 4-4-acetyl-1-piperazinyl phenol, piperazine, 1-acetyl-4-4-hydroxyphenyl, 4-4-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethan-1-one, 1-acetyl-4-4-hydroxylphenyl piperazine
1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethanone

Specifications

Authentic
70% min. (400nm, 1cm cell) (10% min. in N, N-dimethylformamide)
180.0°C to 185.0°C
67914-60-7
100.0
C12H16N2O2
1-acetyl-4-4-hydroxyphenyl piperazine, 4-1-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, n-acetyl-4-4-hydroxyphenyl piperazine, 4-4-acetyl-1-piperazinyl phenol, piperazine, 1-acetyl-4-4-hydroxyphenyl, 4-4-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethan-1-one, 1-acetyl-4-4-hydroxylphenyl piperazine
AGVNLFCRZULMKK-UHFFFAOYSA-N
1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethanone
712441
Powder
1-Acetyl-4-(4-hydroxyphenyl)piperazine, 98%
Glass bottle
Beige or Pink to White
50g
97.5
98%
MFCD00044905
Solubility in water: 4.3g/L (20°C). Other solubilities: 26g/L methanol - 14g/L ethanol, 7g/L 2-propanol - 6g/L chloroform, 6g/L aceton - 140g/L dmf
CC(=O)N1CCN(CC1)C2=CC=C(C=C2)O
220.272
220.27
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 267-744-8