Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetyl-4-(4-hydroxyphenyl)piperazine, 98%

$56.50 - $151.00

Chemical Identifiers

Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
CAS 67914-60-7
Molecular Formula C12H16N2O2
Formula Weight 220.27
MDL Number MFCD00044905
Beilstein 795777
Synonym 4-(4-Acetyl-1-piperazinyl)phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2208314

  SDS
alfa aesar™
B2208314
25g Each for $56.50

AAB2208322

  SDS
alfa aesar™
B2208322
100g Each for $151.00
Description & Specifications

Specifications

Formula Weight 220.27
Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
Assay 98%
Molecular Formula C12H16N2O2
MDL Number MFCD00044905
CAS 67914-60-7
Beilstein 795777
Synonym 4-(4-Acetyl-1-piperazinyl)phenol
Health Hazard 1 Warning
Melting Point 180° to 185°C
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.