Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetyl-4-(4-aminophenyl)piperazine, 97%

$834.74 - $834.74

Chemical Identifiers

Chemical Name or Material 1-Acetyl-4-(4-aminophenyl)piperazine
CAS 92394-00-8
Molecular Formula C12H17N3O
Formula Weight 219.29
MDL Number MFCD01365904
Synonym 4-(4-Acetyl-1-piperazinyl)aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2648606

  SDS
alfa aesar™
H2648606
5g Each for $834.74
Description & Specifications

Specifications

Formula Weight 219.29
Molecular Formula C12H17N3O
MDL Number MFCD01365904
Chemical Name or Material 1-Acetyl-4-(4-aminophenyl)piperazine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 92394-00-8
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Synonym 4-(4-Acetyl-1-piperazinyl)aniline
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.