missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Acetyl-3-indolecarboxaldehyde, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 399700010
Specifications
1-Acetyl-3-indolecarboxaldehyde | |
97.5% min. (GC) | |
C11H9NO2 | |
1g | |
LCJLFGSKHBDOAY-UHFFFAOYSA-N | |
1-acetylindole-3-carbaldehyde | |
89915 | |
98% |
22948-94-3 | |
Glass Bottle | |
MFCD00039691 | |
1-acetyl-1h-indole-3-carbaldehyde, n-acetylindole-3-carboxaldehyde, 1-acetylindole-3-carboxaldehyde, 1-acetyl-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 1-acetyl, 1-acetyl-3-formylindole, zlchem 389, pubchem7227, n-acetylindole-3-aldehyde, acmc-1crl1 | |
CC(=O)N1C=C(C2=CC=CC=C21)C=O | |
187.2 | |
187.2 |
Chemical Identifiers
22948-94-3 | |
187.2 | |
LCJLFGSKHBDOAY-UHFFFAOYSA-N | |
89915 | |
CC(=O)N1C=C(C2=CC=CC=C21)C=O |
C11H9NO2 | |
MFCD00039691 | |
1-acetyl-1h-indole-3-carbaldehyde, n-acetylindole-3-carboxaldehyde, 1-acetylindole-3-carboxaldehyde, 1-acetyl-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 1-acetyl, 1-acetyl-3-formylindole, zlchem 389, pubchem7227, n-acetylindole-3-aldehyde, acmc-1crl1 | |
1-acetylindole-3-carbaldehyde |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 245-347-0
RUO â Research Use Only