missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Acetyl-3-indolecarboxaldehyde, 98%, ACROS Organics™

Chemical Identifiers

CAS 22948-94-3
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.2
MDL Number MFCD00039691
InChI Key LCJLFGSKHBDOAY-UHFFFAOYSA-N
Synonym 1-acetyl-1h-indole-3-carbaldehyde, 1-acetyl-3-formylindole, 1-acetyl-3-indolecarboxaldehyde, 1-acetylindole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 1-acetyl, acmc-1crl1, n-acetylindole-3-aldehyde, n-acetylindole-3-carboxaldehyde, pubchem7227, zlchem 389
PubChem CID 89915
IUPAC Name 1-acetylindole-3-carbaldehyde
SMILES CC(=O)N1C=C(C2=CC=CC=C21)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC399700010
SDS View Documents
Thermo Fisher Scientific
AC399700010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399700050
SDS View Documents
Thermo Fisher Scientific
AC399700050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

22948-94-3
187.2
LCJLFGSKHBDOAY-UHFFFAOYSA-N
89915
CC(=O)N1C=C(C2=CC=CC=C21)C=O
C11H9NO2
MFCD00039691
1-acetyl-1h-indole-3-carbaldehyde, 1-acetyl-3-formylindole, 1-acetyl-3-indolecarboxaldehyde, 1-acetylindole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 1-acetyl, acmc-1crl1, n-acetylindole-3-aldehyde, n-acetylindole-3-carboxaldehyde, pubchem7227, zlchem 389
1-acetylindole-3-carbaldehyde

Specifications

Glass Bottle
1g
97.5% min. (GC)
MFCD00039691
LCJLFGSKHBDOAY-UHFFFAOYSA-N
1-acetylindole-3-carbaldehyde
89915
98%
165°C
22948-94-3
C11H9NO2
1-acetyl-1h-indole-3-carbaldehyde, 1-acetyl-3-formylindole, 1-acetyl-3-indolecarboxaldehyde, 1-acetylindole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 1-acetyl, acmc-1crl1, n-acetylindole-3-aldehyde, n-acetylindole-3-carboxaldehyde, pubchem7227, zlchem 389
CC(=O)N1C=C(C2=CC=CC=C21)C=O
187.2
187.2
1-Acetyl-3-indolecarboxaldehyde
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 245-347-0

SDS