Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetyl-3-ethylaminopyrrolidine, 99%

$82.50 - $82.50

Chemical Identifiers

Chemical Name or Material 1-Acetyl-3-ethylaminopyrrolidine
CAS 916792-58-0
Molecular Formula C8H16N2O
Formula Weight 156.23
MDL Number MFCD08061108
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51001MD

 
alfa aesar™
H51001MD
250mg Each for $82.50
Description & Specifications

Specifications

Formula Weight 156.23
Chemical Name or Material 1-Acetyl-3-ethylaminopyrrolidine
Assay 99%
Molecular Formula C8H16N2O
MDL Number MFCD08061108
CAS 916792-58-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 250mg
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.