Learn More
1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 444440010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester | |
96% min. (GC) | |
C11H17BN2O3 | |
1g | |
GAXOQGDPSCOOLC-UHFFFAOYSA-N | |
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]ethanone | |
17750204 | |
97% |
1150561-76-4 | |
Glass Bottle | |
MFCD09027064 | |
1-acetyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl-ethanone, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl ethanone, 1-acetylpyrazole-4-boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl ethanone, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl ethanone, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl ethan-1-one, ethanone,1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazol-1-yl | |
CC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1 | |
236.08 | |
236.08 |
Chemical Identifiers
1150561-76-4 | |
236.08 | |
GAXOQGDPSCOOLC-UHFFFAOYSA-N | |
17750204 | |
CC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1 |
C11H17BN2O3 | |
MFCD09027064 | |
1-acetyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl-ethanone, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl ethanone, 1-acetylpyrazole-4-boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl ethanone, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl ethanone, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl ethan-1-one, ethanone,1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazol-1-yl | |
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]ethanone |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only