missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444440050

 View more versions of this product

Catalog No. AC444440050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester
Orange to White
5g
96% min. (GC)
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl ethanone, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl ethan-1-one, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl ethanone, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl-ethanone, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl ethanone, 1-acetyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-acetylpyrazole-4-boronic acid pinacol ester, ethanone,1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazol-1-yl
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C(=O)C
236.08
236.08
97%
96% min. consistent with structure (1H-NMR)
47°C to 49°C
1150561-76-4
C11H17BN2O3
GAXOQGDPSCOOLC-UHFFFAOYSA-N
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]ethanone
17750204
Crystalline Powder

Chemical Identifiers

1150561-76-4
236.08
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl ethanone, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl ethan-1-one, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl ethanone, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl-ethanone, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl ethanone, 1-acetyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-acetylpyrazole-4-boronic acid pinacol ester, ethanone,1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazol-1-yl
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]ethanone
C11H17BN2O3
GAXOQGDPSCOOLC-UHFFFAOYSA-N
17750204
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents