missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Acetoxy-4-diethylamino-2-butyne, 98%

Manufacturer:  Alfa Aesar™ H5351114

 View more versions of this product

Catalog No. AAH5351114



Specifications

Specifications

1-Acetoxy-4-diethylamino-2-butyne
1.456
25g
C10H17NO2
MFCD00190224
AQEDWWHCJSXKHP-UHFFFAOYSA-N
4-(diethylamino)but-2-ynyl acetate
2756256
98%
103°C (217°F)
217°C to 218°C
22396-77-6
CH3CO2CH2C≡CCH2N(CH2CH3)2
1-acetoxy-4-diethylamino-2-butyne
CCN(CC)CC#CCOC(=O)C
183.251
183.25

Chemical Identifiers

22396-77-6
183.251
AQEDWWHCJSXKHP-UHFFFAOYSA-N
2756256
CCN(CC)CC#CCOC(=O)C
C10H17NO2
MFCD00190224
1-acetoxy-4-diethylamino-2-butyne
4-(diethylamino)but-2-ynyl acetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents