Please login to your online account to display your discounted pricing

1-Acetamidoadamantane ≥99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  133601000

 View more versions of this product

Catalog No. AC133601000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 193.29
Physical Form Crystalline Powder
Quantity 100g
Color White to yellow
EINECS Number 212-914-9
TSCA TSCA
RTECS Number AB4308000
Melting Point 146° to 149°C
Beilstein 12, 220
CAS Max % 100.0
CAS Min % 99.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
CAS 880-52-4
Molecular Formula C12H19NO
Chemical Name or Material 1-Acetamidoadamantane
Name Note 99+%
Assay Percent Range 99+%
MDL Number MFCD00074730
Infrared Spectrum Authentic
Synonym N-(1-Adamantyl)acetamide

Specifications

Chemical Name or Material 1-Acetamidoadamantane
Name Note 99+%
CAS 880-52-4
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C12H19NO
Formula Weight 193.29
MDL Number MFCD00074730
Synonym N-(1-Adamantyl)acetamide
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents