Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,9-Nonanedithiol, 97%

$46.26 - $46.26

Chemical Identifiers

Chemical Name or Material 1,9-Nonanedithiol
CAS 3489-28-9
Molecular Formula HS(CH2)9SH
Formula Weight 192.39
MDL Number MFCD00004914
Beilstein 1737568
Synonym 1,9-Dimercaptononane; Nonamethylene dimercaptan
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0717203

  SDS
alfa aesar™
L0717203
1g Each for $46.26
Description & Specifications

Specifications

Formula Weight 192.39
Assay 97%
MDL Number MFCD00004914
CAS 3489-28-9
Synonym 1,9-Dimercaptononane; Nonamethylene dimercaptan
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P361-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1,9-Nonanedithiol
TSCA TSCA
Molecular Formula HS(CH2)9SH
Beilstein 1737568
Flash Point >100°C (212°F)
Boiling Point 351° to 352°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P361-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.