missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,9-Nonanedicarboxylic Acid, TCI America™

$206.25 - $693.63

Chemical Identifiers

CAS 1852-04-6
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00004444
InChI Key LWBHHRRTOZQPDM-UHFFFAOYSA-N
Synonym 1,9-nonanedicarboxylic acid, 1,11-undecanedioic acid, undecanedionic acid, 1,9-nonanedicarboxylicacid, undecandioic acid, hendecanedioic acid, unii-7059gfk8nv, nonamethylenedicarboxylic acid, hendecanedioate, undecanedioate
PubChem CID 15816
ChEBI CHEBI:73713
IUPAC Name undecanedioic acid
SMILES C(CCCCC(=O)O)CCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N03335G View Documents TCI America
N03335G
5g Each for $206.25
 
N033325G View Documents TCI America
N033325G
25g Each for $693.63
 
Specifications

Chemical Identifiers

1852-04-6
216.277
LWBHHRRTOZQPDM-UHFFFAOYSA-N
15816
undecanedioic acid
C11H20O4
MFCD00004444
1,9-nonanedicarboxylic acid, 1,11-undecanedioic acid, undecanedionic acid, 1,9-nonanedicarboxylicacid, undecandioic acid, hendecanedioic acid, unii-7059gfk8nv, nonamethylenedicarboxylic acid, hendecanedioate, undecanedioate
CHEBI:73713
C(CCCCC(=O)O)CCCCC(=O)O

Specifications

White-Yellow
5g
C11H20O4
1,9-nonanedicarboxylic acid, 1,11-undecanedioic acid, undecanedionic acid, 1,9-nonanedicarboxylicacid, undecandioic acid, hendecanedioic acid, unii-7059gfk8nv, nonamethylenedicarboxylic acid, hendecanedioate, undecanedioate
C(CCCCC(=O)O)CCCCC(=O)O
216.277
CHEBI:73713
Crystal-Powder at 20°C
1,9-Nonanedicarboxylic Acid
111°C
1852-04-6
MFCD00004444
LWBHHRRTOZQPDM-UHFFFAOYSA-N
undecanedioic acid
15816
216.28
≥97.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0878

TSCA : Yes