Learn More
1,8-Octanedithiol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 310620010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 8-Octanedithiol | |
1191-62-4 | |
100.0 | |
0.9600g/mL | |
>110°C | |
98.5% min. (GC) | |
C8H18S2 | |
HS(CH2)8SH | |
1 g | |
1,8-octanedithiol, 1,8-dimercaptooctane, octamethylene dimercaptan, 1,8-octamethylenedithiol, unii-kh3w3161hs, fema no. 3514, 1,8-octane dithiol, nanothinks tm thio8, acmc-209a1j, dsstox_cid_27474 | |
PGTWZHXOSWQKCY-UHFFFAOYSA-N | |
octane-1,8-dithiol | |
14493 | |
99% |
99% | |
98.5 | |
Colorless to Yellow | |
269.0°C to 270.0°C | |
Authentic | |
Glass bottle | |
1.5030 to 1.5050 | |
MFCD00003574 | |
0.96 | |
Solubility in water: .. Other solubilities: soluble in most organic solvents | |
SCCCCCCCCS | |
178.35 | |
178.36 | |
Liquid |
Chemical Identifiers
1191-62-4 | |
178.35 | |
PGTWZHXOSWQKCY-UHFFFAOYSA-N | |
14493 | |
SCCCCCCCCS |
C8H18S2 | |
MFCD00003574 | |
1,8-octanedithiol, 1,8-dimercaptooctane, octamethylene dimercaptan, 1,8-octamethylenedithiol, unii-kh3w3161hs, fema no. 3514, 1,8-octane dithiol, nanothinks tm thio8, acmc-209a1j, dsstox_cid_27474 | |
octane-1,8-dithiol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physicia
GHS Signal Word: Warning
EINECSNumber : 214-738-8
RTECSNumber : RG9610000
TSCA : TSCA
RUO – Research Use Only