Learn More
1,8-Nonadiyne, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2378414
Description
It is used as starting reagent in the synthesis of 2,6-hexadecadiynoic acid, 2,6-nonadecadiynoic acid and 2,9-hexadecadiynoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as starting reagent in the synthesis of 2,6-hexadecadiynoic acid, 2,6-nonadecadiynoic acid and 2,9-hexadecadiynoic acid.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from heavy metal powders and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.
Specifications
1,8-Nonadiyne | |
-21°C | |
55°C to 56°C (13 mmHg) | |
Characteristic | |
1.449 | |
MFCD00008581 | |
UN3295 | |
1,8-nonadiyne, acmc-1cak0, 1,8-nonadiyne,, 1,8-nonadiyne 6ci, 7ci, 8ci, 9ci | |
DMOVPHYFYSASTC-UHFFFAOYSA-N | |
nona-1,8-diyne | |
16969 | |
97% |
2396-65-8 | |
0.799 | |
41°C (105°F) | |
C9H12 | |
HC≡C(CH2)5C≡CH | |
25 g | |
1733443 | |
Not miscible or difficult to mix in water. | |
C#CCCCCCC#C | |
120.195 | |
120.2 |
Chemical Identifiers
2396-65-8 | |
120.195 | |
DMOVPHYFYSASTC-UHFFFAOYSA-N | |
16969 | |
C#CCCCCCC#C |
C9H12 | |
MFCD00008581 | |
1,8-nonadiyne, acmc-1cak0, 1,8-nonadiyne,, 1,8-nonadiyne 6ci, 7ci, 8ci, 9ci | |
nona-1,8-diyne |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 219-254-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only