missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,8-Nonadiene, 99%

Manufacturer:  Alfa Aesar™ H5350206

 View more versions of this product

Catalog No. AAH5350206



Specifications

Specifications

1,8-Nonadiene
141°C to 142°C
4900-30-5
CH2=CH(CH2)5CH=CH2
UN3295
VJHGSLHHMIELQD-UHFFFAOYSA-N
nona-1,8-diene
78612
99%
26°C (79°F)
5g
C9H16
MFCD00008669
1,8-nonadiene
C=CCCCCCC=C
124.227
124.23

Chemical Identifiers

4900-30-5
124.227
VJHGSLHHMIELQD-UHFFFAOYSA-N
78612
C=CCCCCCC=C
C9H16
MFCD00008669
1,8-nonadiene
nona-1,8-diene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : TSCA

Documents