missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,8-Naphthalic anhydride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 128145000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 8-Naphthalic anhydride | |
81-84-5 | |
100.0 | |
Beige to Brown | |
0.5% max. | |
97% | |
C12H6O3 | |
500 g | |
01,713 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,soluble in acetic acid | |
O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2 | |
198.18 | |
CHEBI:82246 | |
97% |
97% | |
96.0 | |
>269.0°C | |
272°C | |
Authentic | |
Plastic bottle | |
MFCD00006925 | |
17, 521 | |
1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione | |
GRSMWKLPSNHDHA-UHFFFAOYSA-N | |
3-oxatricyclo[7.3.1.0âµ,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione | |
6693 | |
198.18 | |
Powder |
Chemical Identifiers
81-84-5 | |
198.18 | |
GRSMWKLPSNHDHA-UHFFFAOYSA-N | |
6693 | |
3-oxatricyclo[7.3.1.0âµ,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione |
C12H6O3 | |
MFCD00006925 | |
1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione | |
CHEBI:82246 | |
O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2 |
Safety and Handling
MOISTURE SENSITIVE
EINECSNumber : 201-380-2
RTECSNumber : QK5350000
TSCA : TSCA
RUO – Research Use Only