Please login to your online account to display your discounted pricing

1,8-Naphthalaldehydic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 8-Naphthalaldehydic acid
CAS 5811-87-0
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C12H8O3
Formula Weight 200.19g/mol
MDL Number MFCD00134159
Synonym 8-Formyl-1-naphthalenecarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373100050

  SDS
thermo fisher scientific
373100050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color Cream
Melting Point 160° to 167°C
Quantity 5g
Chemical Name or Material 1, 8-Naphthalaldehydic acid
CAS 5811-87-0
CAS Min % 96.0
CAS Max % 100.0
Physical Form Cream powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C12H8O3
Formula Weight 200.19g/mol
MDL Number MFCD00134159
Synonym 8-Formyl-1-naphthalenecarboxylic acid
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.