Please login to your online account to display your discounted pricing

1,8-dimethylnaphthalene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 8-Dimethylnaphthalene
CAS 569-41-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H12
Linear Formula C10H6(CH3)2
Formula Weight 156.23
MDL Number MFCD00004040
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376920010

  SDS
thermo fisher scientific
AC376920010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 156.23
Quantity 1g
Color Off-white to orange-beige
Melting Point 58° to 62°C
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C10H6(CH3)2
CAS 569-41-5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1, 8-Dimethylnaphthalene
Molecular Formula C12H12
MDL Number MFCD00004040
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.