Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,8-Dihydroxy-4,5-dinitroanthraquinone, 97%

$114.66 - $1,473.61

Chemical Identifiers

Chemical Name or Material 1,8-Dihydroxy-4,5-dinitroanthraquinone
CAS 81-55-0
Molecular Formula C14H6N2O8
Formula Weight 330.21
MDL Number MFCD00019149
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2503209

  SDS
alfa aesar™
B2503209
10g Each for $114.66

AAB2503218

  SDS
alfa aesar™
B2503218
50g Each for $409.84

AAB2503230

  SDS
alfa aesar™
B2503230
250g Each for $1,473.61
Description & Specifications

Specifications

Formula Weight 330.21
CAS 81-55-0
Molecular Formula C14H6N2O8
TSCA TSCA
Quantity 10g
Assay 97%
Chemical Name or Material 1,8-Dihydroxy-4,5-dinitroanthraquinone
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Melting Point 225°C (dec.)
MDL Number MFCD00019149
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.