Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane

$232.04 - $232.04

Chemical Identifiers

Chemical Name or Material 1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane
CAS 214078-93-0
Molecular Formula C24H36N4
Formula Weight 380.58
MDL Number MFCD09263312
Synonym N(1),N(8)-Dibenzylcyclam
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26643MD

  SDS
alfa aesar™
H26643MD
250mg Each for $232.04
Description & Specifications

Specifications

Formula Weight 380.58
Chemical Name or Material 1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane
CAS 214078-93-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym N(1),N(8)-Dibenzylcyclam
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Quantity 250mg
MDL Number MFCD09263312
Melting Point 67° to 68°C
Molecular Formula C24H36N4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.