Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,8-Diazabicyclo[5.4.0]undec-7-ene hydrotribromide, 98%

Chemical Identifiers

Chemical Name or Material 1,8-Diazabicyclo[5.4.0]undec-7-ene hydrotribromide
CAS 138666-59-8
Molecular Formula C9H17Br3N2
Formula Weight 392.96
MDL Number MFCD00191982
UN Number UN3261
Synonym DBUHBr_3; 1,8-Diazabicyclo[5.4.0]undec-7-ene hydrobromide perbromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1756903

  SDS
alfa aesar™
L1756903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1756906

  SDS
alfa aesar™
L1756906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 392.96
Chemical Name or Material 1,8-Diazabicyclo[5.4.0]undec-7-ene hydrotribromide
Molecular Formula C9H17Br3N2
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Synonym DBUHBr_3; 1,8-Diazabicyclo[5.4.0]undec-7-ene hydrobromide perbromide
MDL Number MFCD00191982
CAS 138666-59-8
UN Number UN3261
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point 119° to 122°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.