Please login to your online account to display your discounted pricing

1,8-Diazabicyclo[5.4.0]undec-7-ene 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  160610050

 View more versions of this product

Catalog No. AC160610050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 152.24
Quantity 5g
Color Clear colorless to light yellow
Merck Index 15, 2996
TSCA TSCA
Density 1.019g/mL
EINECS Number 229-713-7
Fieser 02,101; 04,119; 05,177; 06,158; 07,87; 11,155; 13,91; 14,109; 15,109; 16,105; 17,99
CAS Max % 100.0
Beilstein 23, V,5, 271
Water 0.5% max. (K.F.)
Refractive Index 1.5210 to 1.5230 (20°C, 589nm)
Synonym 2, 3, 4
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Name Note 98%
CAS 6674-22-2
MDL Number MFCD00006930
Molecular Formula C9H16N2
Assay Percent Range 98%
Chemical Name or Material 1, 8-Diazabicyclo[5.4.0]undec-7-ene

Specifications

Chemical Name or Material 1, 8-Diazabicyclo[5.4.0]undec-7-ene
Name Note 98%
CAS 6674-22-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H16N2
Formula Weight 152.24
MDL Number MFCD00006930
Synonym 2, 3, 4
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents